Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | uncharacterized LOC102800447 | LOC102800447 | Sense | Chr. 2 - 67086446 - 67289244 on NCBI Build GRCh38 | ||
| Downstream | Ewing tumor associated antigen 1 | ETAA1 | ETAA16 | 121.0kb | Sense | Chr. 2 - 67397310 - 67410411 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map