Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | family with sequence similarity 49 member A | FAM49A | -439.0kb | Sense | Chr. 2 - 16549459 - 16665866 on NCBI Build GRCh38 | |
| Downstream | RAD51 associated protein 2 | RAD51AP2 | 403.9kb | Sense | Chr. 2 - 17510584 - 17530414 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map