Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | microRNA 1267 | MIR1267 |
MIRN1267, hsa-mir-1267 |
-55.4kb | Sense | Chr. 13 - 107531171 - 107531248 on NCBI Build GRCh38 |
| Downstream | FAM155A intronic transcript 1 | FAM155A-IT1 | 198.1kb | Sense | Chr. 13 - 107787361 - 107835458 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map