Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | microRNA 548at | MIR548AT | -15.0kb | Sense | Chr. 17 - 42494773 - 42494830 on NCBI Build GRCh38 | |
| Downstream | microRNA 5010 | MIR5010 | mir-5010 | 2.7kb | Sense | Chr. 17 - 42514188 - 42514307 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map