Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | microRNA 1273c | MIR1273C | -397.9kb | Anti-sense | Chr. 6 - 154853360 - 154853436 on NCBI Build GRCh38 | |
| Downstream | claudin 20 | CLDN20 | 10.1kb | Anti-sense | Chr. 6 - 155264013 - 155276548 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map