Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | spastic paraplegia 11 (autosomal recessive) | SPG11 |
ALS5, CMT2X, KIAA1840 |
Anti-sense | Chr. 15 - 44562696 - 44663678 on NCBI Build GRCh38 | |
| Downstream | PAT1 homolog 2 | PATL2 |
Pat1a, hPat1a |
98.4kb | Anti-sense | Chr. 15 - 44665732 - 44711389 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map