Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | C3orf67 antisense RNA 1 | C3orf67-AS1 | Anti-sense | Chr. 3 - 58824471 - 59019093 on NCBI Build GRCh38 | ||
| Downstream | hCG1813818 | LOC339902 | 725.8kb | Sense | Chr. 3 - 59640163 - 59641872 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map