Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | lipopolysaccharide binding protein | LBP | BPIFD2 | -33.3kb | Anti-sense | Chr. 20 - 38346411 - 38377011 on NCBI Build GRCh38 |
| Downstream | small nucleolar RNA host gene 17 | SNHG17 | 8.2kb | Sense | Chr. 20 - 38420592 - 38435375 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map