Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | ribosomal protein L13a pseudogene 20 | RPL13AP20 | RPL13A_9_1211 | -23.6kb | Sense | Chr. 12 - 12875477 - 12876136 on NCBI Build GRCh38 |
| Downstream | microRNA 614 | MIR614 |
MIRN614, hsa-mir-614 |
14.0kb | Sense | Chr. 12 - 12915829 - 12915918 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map