Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | FEZ family zinc finger 2 | FEZF2 |
FEZ, FEZL, FKSG36, TOF, ZFP312, ZNF312 |
-277.3kb | Anti-sense | Chr. 3 - 62369672 - 62373515 on NCBI Build GRCh38 |
| Downstream | keratin 8 pseudogene 35 | KRT8P35 | 443.8kb | Anti-sense | Chr. 3 - 63096246 - 63097755 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map