Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | neuron navigator 1 | NAV1 |
POMFIL3, STEERIN1, UNC53H1 |
Sense | Chr. 1 - 201648322 - 201826974 on NCBI Build GRCh38 | |
| Downstream | IPO9 antisense RNA 1 | IPO9-AS1 | 11.0kb | Anti-sense | Chr. 1 - 201688256 - 201829559 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map