Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | coiled-coil-helix-coiled-coil-helix domain containing 5 | CHCHD5 |
C2orf9, MIC14 |
Sense | Chr. 2 - 112583606 - 112589040 on NCBI Build GRCh38 | |
| Downstream | uncharacterized LOC400999 | FLJ42351 | 55.9kb | Anti-sense | Chr. 2 - 112641830 - 112644180 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map