Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | HHIP antisense RNA 1 | HHIP-AS1 | -40.7kb | Anti-sense | Chr. 4 - 144642916 - 144646019 on NCBI Build GRCh38 | |
| Downstream | anaphase promoting complex subunit 10 | ANAPC10 |
APC10, DOC1 |
278.5kb | Anti-sense | Chr. 4 - 144967117 - 145098541 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map