Assay Details
| Gene Symbol: LOC101929705 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_125864.1 | 1 - 2 | 155 | 89 | ||
| GenBank mRNA | AK024736.1 | - | 1 - 2 | 155 | 89 | |
More Information
Updated Mapping Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0600011898.hg.1 | ENST00000422059.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011898.hg.1 | ENST00000422059.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011898.hg.1 | ENST00000422059.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011898.hg.1 | ENST00000422059.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011898.hg.1 | ENST00000422059.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011898.hg.1 | ENST00000422059.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011898.hg.1 | ENST00000422059.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011898.hg.1 | ENST00000422059.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011898.hg.1 | ENST00000422059.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011898.hg.1 | ENST00000422059.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011898.hg.1 | ENST00000422059.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011898.hg.1 | ENST00000422059.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011898.hg.1 | ENST00000422059.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011898.hg.1 | ENST00000422059.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011898.hg.1 | ENST00000422059.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011898.hg.1 | ENST00000422059.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011898.hg.1 | ENST00000607590.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011898.hg.1 | ENST00000607590.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011898.hg.1 | ENST00000607590.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011898.hg.1 | ENST00000607590.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011898.hg.1 | NR_125864.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011898.hg.1 | NR_125864.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011898.hg.1 | NR_125864.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011898.hg.1 | NR_125864.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011898.hg.1 | NR_125864.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011898.hg.1 | NR_125864.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011898.hg.1 | NR_125864.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011898.hg.1 | NR_125864.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011898.hg.1 | NR_125865.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011898.hg.1 | NR_125865.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011898.hg.1 | NR_125865.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011898.hg.1 | NR_125865.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011898.hg.1 | NR_125865.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011898.hg.1 | NR_125865.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011898.hg.1 | NR_125865.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600181826.hg.1 | ENST00000422059.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600181826.hg.1 | ENST00000422059.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600181826.hg.1 | ENST00000607590.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600181826.hg.1 | NR_125864.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600181826.hg.1 | NR_125865.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600181826.hg.1 | ENST00000422059.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600181826.hg.1 | ENST00000422059.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600181826.hg.1 | ENST00000607590.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600181826.hg.1 | NR_125864.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600181826.hg.1 | NR_125865.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600181826.hg.1 | TC0600011898.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0600181824.hg.1 | ENST00000422059.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600181824.hg.1 | ENST00000422059.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600181824.hg.1 | ENST00000607590.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600181824.hg.1 | NR_125864.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600181824.hg.1 | NR_125865.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600181824.hg.1 | ENST00000422059.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600181824.hg.1 | ENST00000422059.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600181824.hg.1 | ENST00000607590.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600181824.hg.1 | NR_125864.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600181824.hg.1 | NR_125865.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600181824.hg.1 | TC0600011898.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0600181827.hg.1 | ENST00000607590.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600181827.hg.1 | NR_125865.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600181827.hg.1 | ENST00000607590.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600181827.hg.1 | NR_125865.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600181827.hg.1 | TC0600011898.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0600181829.hg.1 | ENST00000422059.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600181829.hg.1 | ENST00000422059.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600181829.hg.1 | NR_125864.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600181829.hg.1 | ENST00000422059.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600181829.hg.1 | ENST00000422059.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600181829.hg.1 | NR_125864.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600181829.hg.1 | TC0600011898.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0600011898.hg.1 | ENST00000422059.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011898.hg.1 | ENST00000422059.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011898.hg.1 | ENST00000422059.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011898.hg.1 | ENST00000422059.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011898.hg.1 | ENST00000422059.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011898.hg.1 | ENST00000422059.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011898.hg.1 | ENST00000422059.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011898.hg.1 | ENST00000422059.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011898.hg.1 | ENST00000422059.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011898.hg.1 | ENST00000422059.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011898.hg.1 | ENST00000422059.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011898.hg.1 | ENST00000422059.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011898.hg.1 | ENST00000422059.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011898.hg.1 | ENST00000422059.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011898.hg.1 | ENST00000422059.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011898.hg.1 | ENST00000422059.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011898.hg.1 | ENST00000607590.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011898.hg.1 | ENST00000607590.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011898.hg.1 | ENST00000607590.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011898.hg.1 | ENST00000607590.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011898.hg.1 | NR_125864.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600011898.hg.1 | NR_125864.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600011898.hg.1 | NR_125864.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600011898.hg.1 | NR_125864.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600011898.hg.1 | NR_125864.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600011898.hg.1 | NR_125864.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600011898.hg.1 | NR_125864.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600011898.hg.1 | NR_125864.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600011898.hg.1 | NR_125865.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600011898.hg.1 | NR_125865.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600011898.hg.1 | NR_125865.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600011898.hg.1 | NR_125865.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600011898.hg.1 | NR_125865.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600011898.hg.1 | NR_125865.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600011898.hg.1 | NR_125865.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600181823.hg.1 | ENST00000422059.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600181823.hg.1 | ENST00000422059.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600181823.hg.1 | ENST00000607590.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600181823.hg.1 | NR_125864.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600181823.hg.1 | NR_125865.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600181823.hg.1 | ENST00000422059.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600181823.hg.1 | ENST00000422059.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600181823.hg.1 | ENST00000607590.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600181823.hg.1 | NR_125864.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600181823.hg.1 | NR_125865.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600181823.hg.1 | TC0600011898.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0600181822.hg.1 | ENST00000422059.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600181822.hg.1 | ENST00000422059.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600181822.hg.1 | NR_125864.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600181822.hg.1 | NR_125865.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600181822.hg.1 | ENST00000422059.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600181822.hg.1 | ENST00000422059.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600181822.hg.1 | NR_125864.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600181822.hg.1 | NR_125865.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600181822.hg.1 | TC0600011898.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0600181825.hg.1 | ENST00000422059.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600181825.hg.1 | ENST00000422059.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600181825.hg.1 | NR_125864.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600181825.hg.1 | ENST00000422059.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600181825.hg.1 | ENST00000422059.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600181825.hg.1 | NR_125864.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600181825.hg.1 | TC0600011898.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0600181821.hg.1 | ENST00000422059.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600181821.hg.1 | ENST00000422059.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600181821.hg.1 | NR_125864.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600181821.hg.1 | NR_125865.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600181821.hg.1 | ENST00000422059.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600181821.hg.1 | ENST00000422059.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600181821.hg.1 | NR_125864.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600181821.hg.1 | NR_125865.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600181821.hg.1 | TC0600011898.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0600181820.hg.1 | ENST00000422059.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600181820.hg.1 | ENST00000422059.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600181820.hg.1 | NR_125864.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600181820.hg.1 | NR_125865.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600181820.hg.1 | ENST00000422059.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600181820.hg.1 | ENST00000422059.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600181820.hg.1 | NR_125864.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600181820.hg.1 | NR_125865.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600181820.hg.1 | TC0600011898.hg.1 | Clariom D TC |

LOC101929705
Genomic Map