Assay Details
| Gene Symbol: COA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017012413.1 | XP_016867902.1 | 5 | 88 | ||
| XM_017012420.1 | XP_016867909.1 | 6 | 88 | |||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | serine/threonine kinase 17a | STK17A | DRAK1 | -12.0kb | Sense | Chr. 7 - 43583093 - 43627379 on NCBI Build GRCh38 |
| Downstream | biliverdin reductase A | BLVRA |
BLVR, BVR, BVRA |
116.9kb | Sense | Chr. 7 - 43758153 - 43807342 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

COA1
Genomic Map