Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | tektin 1 | TEKT1 | -29.3kb | Anti-sense | Chr. 17 - 6799981 - 6831741 on NCBI Build GRCh38 | |
| Downstream | ALOX12 antisense RNA 1 | ALOX12-AS1 | 121.2kb | Anti-sense | Chr. 17 - 6985123 - 7012334 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map