Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | casein kinase 1 alpha 1 like | CSNK1A1L | CK1 | -37.3kb | Sense | Chr. 13 - 37103260 - 37105664 on NCBI Build GRCh38 |
| Downstream | long intergenic non-protein coding RNA 1048 | LINC01048 | 334.7kb | Sense | Chr. 13 - 37481464 - 37484784 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map