Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | toll like receptor 4 | TLR4 |
ARMD10, CD284, TLR-4, TOLL |
-506.2kb | Sense | Chr. 9 - 117704175 - 117717491 on NCBI Build GRCh38 |
| Downstream | BMP/retinoic acid inducible neural specific 1 | BRINP1 |
DBC1, DBCCR1, FAM5A |
940.1kb | Anti-sense | Chr. 9 - 119166630 - 119369461 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map