Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | GMDS antisense RNA 1 (head to head) | GMDS-AS1 | -67.6kb | Sense | Chr. 6 - 2245753 - 2413591 on NCBI Build GRCh38 | |
| Downstream | long intergenic non-protein coding RNA 1600 | LINC01600 |
C6orf195, bA145H9.2 |
138.7kb | Anti-sense | Chr. 6 - 2621913 - 2634603 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map