Assay Details
| Gene Symbol: DCUN1D1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001308101.1 | NP_001295030.1 | 7 | 7062 | 91 | |
| NM_020640.3 | NP_065691.2 | 7 | 7044 | 91 | ||
| XM_011512912.1 | XP_011511214.1 | 7 | 2379 | 91 | ||
| XM_011512914.1 | XP_011511216.1 | 7 | 2418 | 91 | ||
| XM_011512915.2 | XP_011511217.1 | 8 | 2940 | 91 | ||
| XM_011512916.2 | XP_011511218.1 | 7 | 2868 | 91 | ||
| XM_017006633.1 | XP_016862122.1 | 7 | 2190 | 91 | ||
| XM_017006634.1 | XP_016862123.1 | 7 | 2384 | 91 | ||
| GenBank mRNA | AK095773.1 | - | 1 | 1748 | 91 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | ATPase phospholipid transporting 11B (putative) | ATP11B |
ATPIF, ATPIR |
-16.5kb | Anti-sense | Chr. 3 - 182793503 - 182923010 on NCBI Build GRCh38 |
| Downstream | methylcrotonoyl-CoA carboxylase 1 | MCCC1 |
MCC-B, MCCA |
74.5kb | Sense | Chr. 3 - 183015218 - 183099587 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

DCUN1D1
Genomic Map