Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | CELF2 antisense RNA 2 | CELF2-AS2 | -89.2kb | Anti-sense | Chr. 10 - 11071541 - 11105488 on NCBI Build GRCh38 | |
| Downstream | CELF2 antisense RNA 1 | CELF2-AS1 | C10orf31 | 119.8kb | Anti-sense | Chr. 10 - 11316833 - 11344674 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map