Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | EIF1B antisense RNA 1 | EIF1B-AS1 | -2.4kb | Sense | Chr. 3 - 40173145 - 40309698 on NCBI Build GRCh38 | |
| Downstream | ectonucleoside triphosphate diphosphohydrolase 3 | ENTPD3 |
CD39L3, HB6, NTPDase-3 |
73.5kb | Anti-sense | Chr. 3 - 40387156 - 40428626 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map