Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | sorting nexin 13 | SNX13 | RGS-PX1 | -53.0 | Anti-sense | Chr. 7 - 17790761 - 17940518 on NCBI Build GRCh38 |
| Downstream | phosphoribosyl pyrophosphate synthetase 1-like 1 | PRPS1L1 |
PRPS1, PRPS3, PRPSL, PRS-III |
83.9kb | Anti-sense | Chr. 7 - 18026774 - 18027863 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map