Assay Details
| Gene Symbol: LOC100291105 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK096426.1 | - | 1 - 2 | 789 | 64 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | RNA binding motif protein 38 | RBM38 |
HSRNASEB, RNPC1, SEB4B, SEB4D, dJ800J21.2 |
Anti-sense | Chr. 20 - 57391398 - 57409333 on NCBI Build GRCh38 | |
| Downstream | CCCTC-binding factor like | CTCFL |
BORIS, CT27, CTCF-T, HMGB1L1, dJ579F20.2 |
102.9kb | Sense | Chr. 20 - 57495965 - 57525652 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

LOC100291105
Genomic Map