Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | microRNA 1265 | MIR1265 |
MIRN1265, hsa-mir-1265 |
-87.8kb | Anti-sense | Chr. 10 - 14436576 - 14436661 on NCBI Build GRCh38 |
| Downstream | cerebral dopamine neurotrophic factor | CDNF | ARMETL1 | 291.4kb | Sense | Chr. 10 - 14819245 - 14838073 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map