Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | NADPH oxidase organizer 1 | NOXO1 |
P41NOX, P41NOXA, P41NOXB, P41NOXC, SH3PXD5, SNX28 |
Anti-sense | Chr. 16 - 1978917 - 1982751 on NCBI Build GRCh38 | |
| Downstream |
growth factor, augmenter of liver regeneration |
GFER |
ALR, ERV1, HERV1, HPO, HPO1, HPO2, HSS |
2.4kb | Sense | Chr. 16 - 1984149 - 1987749 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map