Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | metallothionein 1 pseudogene 3 | MT1P3 |
C20orf127, MTL4, dJ614O4.6 |
-44.0kb | Anti-sense | Chr. 20 - 35218009 - 35218185 on NCBI Build GRCh38 |
| Downstream | MMP24 antisense RNA 1 | MMP24-AS1 | 12.1kb | Sense | Chr. 20 - 35276333 - 35278157 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map