Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
protein tyrosine phosphatase, non-receptor type 13 |
PTPN13 |
FAP-1, PNP1, PTP-BAS, PTP-BL, PTP1E, PTPL1, PTPLE, hPTP1E |
Sense | Chr. 4 - 86594307 - 86815179 on NCBI Build GRCh38 | |
| Downstream | solute carrier family 10 member 6 | SLC10A6 | SOAT | 29.8kb | Anti-sense | Chr. 4 - 86823468 - 86849263 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map