Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | ras homolog family member J | RHOJ |
ARHJ, RASL7B, TC10B, TCL |
Sense | Chr. 14 - 63204127 - 63293512 on NCBI Build GRCh38 | |
| Downstream | glycoprotein hormone beta 5 | GPHB5 |
B5, GPB5, ZLUT1 |
23.6kb | Anti-sense | Chr. 14 - 63312835 - 63318879 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map