Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | Rho GTPase activating protein 15 | ARHGAP15 | BM046 | Sense | Chr. 2 - 143070973 - 143768352 on NCBI Build GRCh38 | |
| Downstream | uncharacterized LOC101928361 | LOC101928361 | 272.7kb | Sense | Chr. 2 - 143598906 - 143608759 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map