Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | ubiquitin like modifier activating enzyme 3 | UBA3 |
NAE2, UBE1C, hUBA3 |
Sense | Chr. 3 - 69054730 - 69080373 on NCBI Build GRCh38 | |
| Downstream | ADP ribosylation factor like GTPase 6 interacting protein 5 | ARL6IP5 |
DERP11, GTRAP3-18, HSPC127, JWA, PRAF3, Yip6b, addicsin, hp22, jmx |
23.9kb | Anti-sense | Chr. 3 - 69084939 - 69106088 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map