Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | transmembrane protein 30B | TMEM30B | CDC50B | -58.7kb | Anti-sense | Chr. 14 - 61277370 - 61281812 on NCBI Build GRCh38 |
| Downstream | uncharacterized LOC101927780 | LOC101927780 | 212.9kb | Anti-sense | Chr. 14 - 61556272 - 61570653 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map