Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | uncharacterized LOC153910 | LOC153910 | -117.1kb | Sense | Chr. 6 - 142526455 - 142637889 on NCBI Build GRCh38 | |
| Downstream | long intergenic non-protein coding RNA 1277 | LINC01277 | 208.6kb | Sense | Chr. 6 - 142966421 - 143037582 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map