Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | phenylalanine hydroxylase | PAH |
PH, PKU, PKU1 |
-10.3kb | Sense | Chr. 12 - 102838326 - 102917603 on NCBI Build GRCh38 |
| Downstream | achaete-scute family bHLH transcription factor 1 | ASCL1 |
ASH1, HASH1, MASH1, bHLHa46 |
27.9kb | Anti-sense | Chr. 12 - 102957674 - 102960516 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map