Assay Details
| Gene Symbol: LOC101927078 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_130785.1 | 5 - 6 | 560 | 64 | ||
| GenBank mRNA | AK097686.1 | - | 5 - 6 | 560 | 64 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | YTH domain containing 2 | YTHDC2 | CAHL | -890.2kb | Anti-sense | Chr. 5 - 113513683 - 113595287 on NCBI Build GRCh38 |
| Downstream | uncharacterized LOC101927059 | LOC101927059 | 87.0kb | Anti-sense | Chr. 5 - 114576041 - 114579970 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
Set Membership: |
|

LOC101927078
Genomic Map