| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01010041.hg.1 |
NR_038403.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01010041.hg.1 |
TC01000589.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01010042.hg.1 |
NR_038403.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01010042.hg.1 |
TC01000589.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01010044.hg.1 |
NR_038403.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01010044.hg.1 |
TC01000589.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01010043.hg.1 |
NR_038403.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01010043.hg.1 |
TC01000589.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01010040.hg.1 |
NR_038403.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01010040.hg.1 |
TC01000589.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008166.hg.1 |
ENST00000602433.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008166.hg.1 |
ENST00000602433.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008166.hg.1 |
ENST00000602433.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008166.hg.1 |
ENST00000602433.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008166.hg.1 |
ENST00000602433.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008166.hg.1 |
ENST00000602433.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008166.hg.1 |
ENST00000602433.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008166.hg.1 |
ENST00000602433.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008166.hg.1 |
NR_038403.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008166.hg.1 |
NR_038403.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008166.hg.1 |
NR_038403.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008166.hg.1 |
NR_038403.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008166.hg.1 |
NR_038403.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008166.hg.1 |
NR_038403.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008166.hg.1 |
NR_038403.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008166.hg.1 |
NR_038403.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01010039.hg.1 |
NR_038403.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01010039.hg.1 |
TC01000589.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01010046.hg.1 |
NR_038403.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01010046.hg.1 |
TC01000589.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR0100158460.hg.1 |
ENST00000602433.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100158460.hg.1 |
NR_038403.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100158460.hg.1 |
ENST00000602433.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100158460.hg.1 |
NR_038403.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100158460.hg.1 |
TC0100008166.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0100158461.hg.1 |
ENST00000602433.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100158461.hg.1 |
NR_038403.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100158461.hg.1 |
ENST00000602433.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100158461.hg.1 |
NR_038403.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100158461.hg.1 |
TC0100008166.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0100008166.hg.1 |
ENST00000602433.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100008166.hg.1 |
ENST00000602433.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100008166.hg.1 |
ENST00000602433.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100008166.hg.1 |
ENST00000602433.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100008166.hg.1 |
ENST00000602433.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100008166.hg.1 |
ENST00000602433.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100008166.hg.1 |
ENST00000602433.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100008166.hg.1 |
ENST00000602433.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100008166.hg.1 |
NR_038403.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100008166.hg.1 |
NR_038403.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100008166.hg.1 |
NR_038403.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100008166.hg.1 |
NR_038403.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100008166.hg.1 |
NR_038403.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100008166.hg.1 |
NR_038403.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100008166.hg.1 |
NR_038403.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100008166.hg.1 |
NR_038403.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100158463.hg.1 |
ENST00000602433.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100158463.hg.1 |
NR_038403.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100158463.hg.1 |
ENST00000602433.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100158463.hg.1 |
NR_038403.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100158463.hg.1 |
TC0100008166.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0100158459.hg.1 |
ENST00000602433.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100158459.hg.1 |
NR_038403.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100158459.hg.1 |
ENST00000602433.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100158459.hg.1 |
NR_038403.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100158459.hg.1 |
TC0100008166.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0100158462.hg.1 |
ENST00000602433.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100158462.hg.1 |
NR_038403.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100158462.hg.1 |
ENST00000602433.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100158462.hg.1 |
NR_038403.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100158462.hg.1 |
TC0100008166.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0100158465.hg.1 |
ENST00000602433.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100158465.hg.1 |
NR_038403.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100158465.hg.1 |
ENST00000602433.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100158465.hg.1 |
NR_038403.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100158465.hg.1 |
TC0100008166.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0100158458.hg.1 |
ENST00000602433.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100158458.hg.1 |
NR_038403.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100158458.hg.1 |
ENST00000602433.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100158458.hg.1 |
NR_038403.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100158458.hg.1 |
TC0100008166.hg.1 |
Clariom D TC |