Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | LYR motif containing 9 | LYRM9 |
C17orf108, HSD24 |
Anti-sense | Chr. 17 - 27878314 - 27893383 on NCBI Build GRCh38 | |
| Downstream | uncharacterized LOC105371708 | LOC105371708 | 49.2kb | Anti-sense | Chr. 17 - 27928924 - 27991731 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

Genomic Map