Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | ST6 beta-galactoside alpha-2,6-sialyltransferase 2 | ST6GAL2 |
SIAT2, ST6GalII |
-646.7kb | Sense | Chr. 2 - 106801600 - 106887410 on NCBI Build GRCh38 |
| Downstream | gastric cancer associated transcript 1 (non-protein coding) | GACAT1 | LINC00876 | 218.6kb | Anti-sense | Chr. 2 - 107754112 - 107822129 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map