Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | microRNA 4722 | MIR4722 | -2.3kb | Anti-sense | Chr. 16 - 88716278 - 88716337 on NCBI Build GRCh38 | |
| Downstream | uncharacterized LOC100289580 | LOC100289580 | 10.7kb | Sense | Chr. 16 - 88731180 - 88741425 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map