Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | synaptotagmin like 3 | SYTL3 | SLP3 | -26.9kb | Anti-sense | Chr. 6 - 158644881 - 158766298 on NCBI Build GRCh38 |
| Downstream | EZR antisense RNA 1 | EZR-AS1 | 24.1kb | Anti-sense | Chr. 6 - 158817979 - 158822239 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map