Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | microRNA 4304 | MIR4304 | -10.8kb | Sense | Chr. 12 - 123010667 - 123010728 on NCBI Build GRCh38 | |
| Downstream | uncharacterized LOC100507091 | LOC100507091 | 57.6kb | Anti-sense | Chr. 12 - 123081384 - 123084744 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map