Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | myelin protein zero like 2 | MPZL2 |
EVA, EVA1 |
-57.0 | Anti-sense | Chr. 11 - 118253416 - 118264536 on NCBI Build GRCh38 |
| Downstream | CD3e molecule | CD3E |
IMD18, T3E, TCRE |
37.8kb | Sense | Chr. 11 - 118304580 - 118316175 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map