Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | uncharacterized LOC105378098 | LOC105378098 | -801.2kb | Anti-sense | Chr. 6 - 161470977 - 161484045 on NCBI Build GRCh38 | |
| Downstream | PARK2 coregulated | PACRG |
GLUP, HAK005771, PARK2CRG |
439.4kb | Anti-sense | Chr. 6 - 162726870 - 163315492 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map