Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | long intergenic non-protein coding RNA 1214 | LINC01214 | -105.7kb | Anti-sense | Chr. 3 - 150265407 - 150323747 on NCBI Build GRCh38 | |
| Downstream | stress-associated endoplasmic reticulum protein 1 | SERP1 | RAMP4 | 112.3kb | Anti-sense | Chr. 3 - 150541993 - 150603177 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map