Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | long intergenic non-protein coding RNA 1260 | LINC01260 | -34.6kb | Sense | Chr. 20 - 44656451 - 44671739 on NCBI Build GRCh38 | |
| Downstream | WNT1 inducible signaling pathway protein 2 | WISP2 |
CCN5, CT58, CTGF-L |
8.2kb | Anti-sense | Chr. 20 - 44714204 - 44728041 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

Genomic Map