Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | microRNA 183 | MIR183 |
MIRN183, miR-183, miRNA183 |
-199.0 | Anti-sense | Chr. 7 - 129774905 - 129775014 on NCBI Build GRCh38 |
| Downstream | ubiquitin conjugating enzyme E2 H | UBE2H |
E2-20K, GID3, UBC8, UBCH, UBCH2 |
54.3kb | Anti-sense | Chr. 7 - 129830732 - 129952960 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map