| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800006937.hg.1 |
ENST00000363023.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800006937.hg.1 |
ENST00000523103.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800006937.hg.1 |
ENST00000523103.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800006937.hg.1 |
ENST00000523103.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800006937.hg.1 |
ENST00000523103.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800006937.hg.1 |
ENST00000523103.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800006937.hg.1 |
NR_047509.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800006937.hg.1 |
NR_047509.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800006937.hg.1 |
NR_047509.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800006937.hg.1 |
NR_047509.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800006937.hg.1 |
NR_047509.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0800147021.hg.1 |
ENST00000523103.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800147021.hg.1 |
NR_047509.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0800147021.hg.1 |
ENST00000523103.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800147021.hg.1 |
NR_047509.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0800147021.hg.1 |
TC0800006937.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0800006937.hg.1 |
ENST00000363023.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800006937.hg.1 |
ENST00000523103.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800006937.hg.1 |
ENST00000523103.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800006937.hg.1 |
ENST00000523103.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800006937.hg.1 |
ENST00000523103.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800006937.hg.1 |
ENST00000523103.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800006937.hg.1 |
NR_047509.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800006937.hg.1 |
NR_047509.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800006937.hg.1 |
NR_047509.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800006937.hg.1 |
NR_047509.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800006937.hg.1 |
NR_047509.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0800147020.hg.1 |
ENST00000363023.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800147020.hg.1 |
ENST00000523103.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800147020.hg.1 |
NR_047509.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0800147020.hg.1 |
ENST00000363023.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800147020.hg.1 |
ENST00000523103.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800147020.hg.1 |
NR_047509.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0800147020.hg.1 |
TC0800006937.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0800147019.hg.1 |
ENST00000523103.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800147019.hg.1 |
NR_047509.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0800147019.hg.1 |
ENST00000523103.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800147019.hg.1 |
NR_047509.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0800147019.hg.1 |
TC0800006937.hg.1 |
Clariom D TC |