Assay Details
| Gene Symbol: LINGO1-AS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_120361.1 | 1 - 2 | 134 | 90 | ||
| GenBank mRNA | BC038758.1 | - | 1 - 2 | 133 | 90 | |
| BI463387.1 | - | 1 - 2 | 138 | 90 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | leucine rich repeat and Ig domain containing 1 | LINGO1 |
LERN1, LRRN6A, UNQ201 |
Anti-sense | Chr. 15 - 77613024 - 77820900 on NCBI Build GRCh38 | |
| Downstream | golgin A6 family member A pseudogene | LOC645752 | 247.1kb | Anti-sense | Chr. 15 - 77914217 - 77926846 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

LINGO1-AS2
Genomic Map