Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | GIT ArfGAP 2 | GIT2 |
CAT-2, CAT2, PKL |
-1.0kb | Sense | Chr. 12 - 109929792 - 109996389 on NCBI Build GRCh38 |
| Downstream | ankyrin repeat domain 13A | ANKRD13A |
ANKRD13, NY-REN-25 |
42.0 | Anti-sense | Chr. 12 - 109999169 - 110039432 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map