Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | bromodomain adjacent to zinc finger domain 1A | BAZ1A |
ACF1, WALp1, WCRF180, hACF1 |
Anti-sense | Chr. 14 - 34752731 - 34875647 on NCBI Build GRCh38 | |
| Downstream | immunoglobulin (CD79A) binding protein 1 pseudogene 1 | IGBP1P1 | C14orf19 | 63.5kb | Sense | Chr. 14 - 34939922 - 34940496 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map